missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroindole, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 311922500

 View more versions of this product

Catalog No. AC311922500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

5-Fluoroindole
399-52-0
100.0
Authentic
Glass bottle
MFCD00005671
5-fluoroindole, 1h-indole, 5-fluoro, 5-fluoro indole, indole, 5-fluoro, 5-fluoro-indole, 5-fluor-1h-indol, 5-fluoroindol, 5-fluoro-indol, 3fuf, pubchem1681
FC1=CC=C2NC=CC2=C1
135.14
CHEBI:72818
98%
98%
97.5
>110°C
97.5% min. (GC)
C8H6FN
250mg
ODFFPRGJZRXNHZ-UHFFFAOYSA-N
5-fluoro-1H-indole
67861
135.14

Chemical Identifiers

399-52-0
135.14
ODFFPRGJZRXNHZ-UHFFFAOYSA-N
67861
5-fluoro-1H-indole
C8H6FN
MFCD00005671
5-fluoroindole, 1h-indole, 5-fluoro, 5-fluoro indole, indole, 5-fluoro, 5-fluoro-indole, 5-fluor-1h-indol, 5-fluoroindol, 5-fluoro-indol, 3fuf, pubchem1681
CHEBI:72818
FC1=CC=C2NC=CC2=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 206-917-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only