missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroindole-3-carboxaldehyde, 98%

Catalog No. AAL17507MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL17507MD 250 mg
AAL1750703 1 g
AAL1750706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL17507MD Supplier Thermo Scientific Chemicals Supplier No. L17507MD

CAS: 2338-71-8 | C9H6FNO | 163.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2338-71-8
Molecular Formula C9H6FNO
Molecular Weight (g/mol) 163.15
MDL Number MFCD00022719
InChI Key YUAJKGBLPVLADK-UHFFFAOYSA-N
Synonym 5-fluoroindole-3-carboxaldehyde, 5-fluoro-1h-indole-3-carboxaldehyde, 5-fluoroindole-3-carbaldehyde, 1h-indole-3-carboxaldehyde, 5-fluoro, 3-formyl-5-fluoroindole, pubchem7267, zlchem 1197, 5-fluoro-3-formylindole, 5-fluoroindole-3-aldehyde, acmc-209g2v
PubChem CID 259089
IUPAC Name 5-fluoro-1H-indole-3-carbaldehyde
SMILES FC1=CC=C2NC=C(C=O)C2=C1

Specifications

Melting Point 164°C
Quantity 250 mg
Beilstein 1449216
Sensitivity Air Sensitive
Formula Weight 163.15
Percent Purity 98%
Chemical Name or Material 5-Fluoroindole-3-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only