missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorobenzo[b]thiophene-2-carboxylic acid, 96%

Catalog No. AAH3323903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3323903 1 g
AAH3323906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3323903 Supplier Thermo Scientific Chemicals Supplier No. H3323903

CAS: 70060-13-8 | C9H5FO2S | 196.195 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70060-13-8
Molecular Formula C9H5FO2S
Molecular Weight (g/mol) 196.195
MDL Number MFCD01927184
InChI Key PLVPMOSTGNZKQQ-UHFFFAOYSA-N
Synonym 5-fluorobenzo b thiophene-2-carboxylic acid, 5-fluoro-benzo b thiophene-2-carboxylic acid, benzo b thiophene-2-carboxylicacid, 5-fluoro, 2-carboxy-5-fluoro-1-benzothiophene, 5-fluoro-2-benzo b thiophenecarboxylic acid, 5-fluoro-2-benzo-b thiophenecarboxylic acid, 5-fluorobenzothiophene-2-carboxylic acid, benzo b thiophene-2-carboxylic acid, 5-fluoro
PubChem CID 901439
IUPAC Name 5-fluoro-1-benzothiophene-2-carboxylic acid
SMILES C1=CC2=C(C=C1F)C=C(S2)C(=O)O

Specifications

Melting Point 257°C to 262°C
Quantity 1 g
Formula Weight 196.19
Percent Purity 96%
Chemical Name or Material 5-Fluorobenzo[b]thiophene-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only