missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-3-methylindole 98.0+%, TCI America™

Catalog No. F1053200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1053200MG 200 mg
F10531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F1053200MG Supplier TCI America Supplier No. F1053200MG

Chemical Identifiers

CAS 392-13-2
Molecular Formula C9H8FN
Molecular Weight (g/mol) 149.17
MDL Number MFCD03095377
InChI Key CSDHAGJNOQIBHZ-UHFFFAOYSA-N
PubChem CID 22351503
IUPAC Name 5-fluoro-3-methyl-1H-indole
SMILES CC1=CNC2=CC=C(F)C=C12

Specifications

Melting Point 85°C
Quantity 200 mg
Formula Weight 149.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-3-methylindole