missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride, 96%

Catalog No. p-7031828
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3304403 1 g
AAH3304406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3304403 Supplier Thermo Scientific Chemicals Supplier No. H3304403

CAS: 84163-16-6 | C12H14ClFN2O | 256.705 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84163-16-6
Molecular Formula C12H14ClFN2O
Molecular Weight (g/mol) 256.705
MDL Number MFCD06200879
InChI Key HSOJFILHZZEWJT-UHFFFAOYSA-N
Synonym 5-fluoro-3-piperidin-4-yl benzo d isoxazole hydrochloride, 5-fluoro-3-piperidin-4-yl-1,2-benzisoxazole hydrochloride, 5-fluoro-3-4-piperidinyl-1,2-benzisoxazole hydrochloride, 5-fluoro-3-piperidin-4-yl-1,2-benzoxazole hydrochloride, 5-fluoro-3-piperidin-4-yl benzo d isoxazole hcl, 4-5-fluoro-1,2-benzisoxazol-3-yl piperidine hydrochloride, 1,2-benzisoxazole,5-fluoro-3-4-piperidinyl-, hydrochloride, pubchem11469, 5-fluoro-3-piperidin-4-yl benzisoxazole hcl salt, 5-fluoro-3-4-piperidyl benzo d isoxazole, chloride
PubChem CID 2794754
IUPAC Name 5-fluoro-3-piperidin-4-yl-1,2-benzoxazole;hydrochloride
SMILES C1CNCCC1C2=NOC3=C2C=C(C=C3)F.Cl

Specifications

Melting Point ∽298°C (decomposition)
Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 256.71
Percent Purity 96%
Chemical Name or Material 5-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only