missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-nitroaniline 98.0+%, TCI America™

Catalog No. p-7129900
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07895G 5 g
F078925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07895G Supplier TCI America Supplier No. F07895G
Only null left

Chemical Identifiers

CAS 2369-11-1
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.12
MDL Number MFCD00034065
InChI Key PEDMFCHWOVJDNW-UHFFFAOYSA-N
Synonym 2-nitro-5-fluoroaniline, 5-fluoro-2-nitrophenylamine, 2-amino-4-fluoronitrobenzene, 3-fluoro-6-nitroaniline, 5-fluoro-2-nitrobenzenamine, benzenamine, 5-fluoro-2-nitro, pubchem4476, acmc-1cegf, 5-fluoro-2-nitro-aniline, intermediates-zcf02632
PubChem CID 75399
IUPAC Name 5-fluoro-2-nitroaniline
SMILES C1=CC(=C(C=C1F)N)[N+](=O)[O-]

Specifications

Melting Point 98°C
Quantity 5 g
Formula Weight 156.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-2-nitroaniline