missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-nitroaniline, 97%

Catalog No. p-7053986
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6180606 5 g
AAH6180614 25 g
AAH6180622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6180606 Supplier Thermo Scientific Chemicals Supplier No. H6180606
Only null left

CAS: 2369-11-1 | C6H5FN2O2 | 156.12 g/mol

5-Fluoro-2-nitroaniline as a intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluoro-2-nitroaniline as a intermediate in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 2369-11-1
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.12
MDL Number MFCD00034065
InChI Key PEDMFCHWOVJDNW-UHFFFAOYSA-N
IUPAC Name 5-fluoro-2-nitroaniline

Specifications

Quantity 5 g
UN Number UN2811
Synonym 4-bromo-2,6-xylenol, phenol, 4-bromo-2,6-dimethyl, 2,6-dimethyl-4-bromophenol, 2,6-xylenol, 4-bromo, pubchem9579, 2, 4-bromo, acmc-2097aq, 4-bromo-2,6 dimethylphenol, 5-bromo-2-hydroxy-m-xylene, ksc495g0d
Solubility Information Slightly soluble in water.
SMILES NC1=CC(F)=CC=C1[N+]([O-])=O
PubChem CID 16919
Formula Weight 156.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Fluoro-2-nitroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only