missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. f07755g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F07755G Supplier TCI America Supplier No. F07755G
Only null left

Chemical Identifiers

CAS 179897-94-0
Molecular Formula C7H8BFO3
MDL Number MFCD01863526
InChI Key CCQKIRUMTHHPSX-UHFFFAOYSA-N
PubChem CID 2782673
IUPAC Name (5-fluoro-2-methoxyphenyl)boronic acid

Specifications

Melting Point 158°C
Color White-Yellow
Quantity 5 g
Synonym 5-fluoro-2-methoxyphenyl boronic acid, 2-methoxy-5-fluorophenylboronic acid, 5-fluoro-2-methoxybenzeneboronic acid, 2-methoxy-5-fluorobenzeneboronic acid, 5-fluoro-2-methoxyphenylboronicacid, 2-borono-4-fluoroanisole, 3-fluoro-6-methoxybenzeneboronic acid, 5-fluoro-2-methoxyphenyl boranediol, 4-fluoroanisole-2-boronic acid, boronic acid, 5-fluoro-2-methoxyphenyl
SMILES COC1=CC=C(F)C=C1B(O)O
Molecular Weight (g/mol) 169.95
Formula Weight 169.95
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride)