missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-hydroxypyridine, 97%

Catalog No. p-4228678
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432580050 5 g Glass bottle
AC432580250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432580050 Supplier Thermo Scientific Chemicals Supplier No. 432580050

CAS: 51173-05-8 | C5H4FNO | 113.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51173-05-8
Molecular Formula C5H4FNO
Molecular Weight (g/mol) 113.09
MDL Number MFCD03092918
InChI Key KLULSYPVWLJZAO-UHFFFAOYSA-N
Synonym 5-fluoro-2-hydroxypyridine, 5-fluoropyridin-2-ol, 2-hydroxy-5-fluoropyridine, 5-fluoropyridin-2 1h-one, 3-fluoro-6-hydroxypyridine, 2 1h-pyridinone, 5-fluoro, 5-fluoro-2 1h-pyridinone, 5-fluoro-2-pyridinol, 5-fluoropyridin-2-ol hydrobromide, pubchem6566
PubChem CID 2762864
IUPAC Name 5-fluoro-1H-pyridin-2-one
SMILES C1=CC(=O)NC=C1F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 152.0°C to 153.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 113.09
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-2-hydroxypyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
If medical advice is needed,have product container or label at hand.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only