missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ 5-FAM, SE (5-Carboxyfluorescein, Succinimidyl Ester), single isomer
Description
Searching for superior alternatives to fluorescein? Our Alexa Fluor Dye Series offers everything you're looking for and more.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | 5-FAM SE |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Amine |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Active Ester, Succinimidyl Ester |
| Label or Dye | FITC (Fluorescein) |
| Label Type | Classic Dyes |
For Research Use Only. Not for use in diagnostic procedures.