missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 5'-Ethynyl-2'-deoxyuridine

Catalog No. AC466382500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466382500 250 mg
AC466380010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466382500 Supplier Thermo Scientific Chemicals Supplier No. 466382500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

252.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Molecular Weight (g/mol) 252.23
Product Type 5 '-Ethynyl-2 '-deoxyuridine
Quantity 250 mg
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in dmso, methanol

RUO – Research Use Only