missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Ethylidene-2-norbornene 98.0+%, TCI America™
$41.02 - $123.03
Chemical Identifiers
| CAS | 16219-75-3 |
|---|---|
| Molecular Formula | C9H12 |
| Molecular Weight (g/mol) | 120.195 |
| MDL Number | MFCD00167576 |
| InChI Key | OJOWICOBYCXEKR-WHZSQGQPSA-N |
| Synonym | 5-ethylidene-2-norbornene, unii-85015b2v0r component, unii-q4d40uvr0r component |
| PubChem CID | 12456033 |
| IUPAC Name | (1R,4R,5E)-5-ethylidenebicyclo[2.2.1]hept-2-ene |
| SMILES | CC=C1CC2CC1C=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E024425ML
|
TCI America
E024425ML |
25 mL |
Each for $41.02
|
|
|||||
|
E0244500ML
|
TCI America
E0244500ML |
500 mL |
Each for $123.03
|
|
|||||
Chemical Identifiers
| 16219-75-3 | |
| 120.195 | |
| OJOWICOBYCXEKR-WHZSQGQPSA-N | |
| 12456033 | |
| CC=C1CC2CC1C=C2 |
| C9H12 | |
| MFCD00167576 | |
| 5-ethylidene-2-norbornene, unii-85015b2v0r component, unii-q4d40uvr0r component | |
| (1R,4R,5E)-5-ethylidenebicyclo[2.2.1]hept-2-ene |
Specifications
| 16219-75-3 | |
| 147°C | |
| MFCD00167576 | |
| 3295 | |
| OJOWICOBYCXEKR-WHZSQGQPSA-N | |
| (1R,4R,5E)-5-ethylidenebicyclo[2.2.1]hept-2-ene | |
| 12456033 | |
| ≥98.0% (GC) | |
| 5-Ethylidene-2-norbornene |
| Yellow | |
| C9H12 | |
| 25 mL | |
| 5-ethylidene-2-norbornene, unii-85015b2v0r component, unii-q4d40uvr0r component | |
| CC=C1CC2CC1C=C2 | |
| 120.195 | |
| 120.20 | |
| Liquid |
Safety and Handling
EINECSNumber : (4)-0602
RTECSNumber : RB9450000
TSCA : Yes