missing translation for 'onlineSavingsMsg'
Learn More

5-Ethylidene-2-norbornene 98.0+%, TCI America™

Catalog No. E024425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E024425ML 25 mL
E0244500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E024425ML Supplier TCI America Supplier No. E024425ML

Chemical Identifiers

CAS 16219-75-3
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00167576
InChI Key OJOWICOBYCXEKR-WHZSQGQPSA-N
Synonym 5-ethylidene-2-norbornene, unii-85015b2v0r component, unii-q4d40uvr0r component
PubChem CID 12456033
IUPAC Name (1R,4R,5E)-5-ethylidenebicyclo[2.2.1]hept-2-ene
SMILES CC=C1CC2CC1C=C2

Specifications

Color Yellow
Boiling Point 147°C
Quantity 25 mL
UN Number 3295
Formula Weight 120.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Ethylidene-2-norbornene
EINECSNumber (4)-0602
RTECSNumber RB9450000
TSCA Yes