missing translation for 'onlineSavingsMsg'
Learn More

5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone 97.0+%, TCI America™

Catalog No. p-7129157
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08005G 5 g
E080025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08005G Supplier TCI America Supplier No. E08005G
Only null left

Chemical Identifiers

CAS 698-10-2
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD00036673
InChI Key IUFQZPBIRYFPFD-UHFFFAOYSA-N
Synonym 5-ethyl-3-hydroxy-4-methyl-2 5h-furanone, 5-ethyl-3-hydroxy-4-methylfuran-2 5h-one, 2 5h-furanone, 5-ethyl-3-hydroxy-4-methyl, 3-hydroxy-4-methyl-5-ethyl-2-furanone, fema no. 3153, 4-methyl-5-ethyl-3-hydroxyfuranone, ethyl fenugreek lactone, 3-hydroxy-4-methyl-5-ethyl-2 5h furanone, 5-ethyl-3-hydroxy-4-methyl-2 5h furanone, 2-hydroxy-3-methyl-gamma-2-hexenolactone
PubChem CID 61199
ChEBI CHEBI:67940
IUPAC Name 2-ethyl-4-hydroxy-3-methyl-2H-furan-5-one
SMILES CCC1C(=C(C(=O)O1)O)C

Specifications

Color White-Yellow
Boiling Point 94°C
Quantity 5 g
Formula Weight 142.15
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone