missing translation for 'onlineSavingsMsg'
Learn More

5-Ethyl-2-pyridineethanol 98.0+%, TCI America™

Catalog No. p-7129069
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
E05625G 5 g
E056225G 25 g
2 options

Catalog No. E05625G

Supplier: TCI America E05625G

Only null left

Chemical Identifiers

CAS 5223-06-3
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00129040
InChI Key OUJMXIPHUCDRAS-UHFFFAOYSA-N
Synonym 5-ethyl-2-pyridineethanol, 2-5-ethylpyridin-2-yl ethanol, 2-5-ethyl-2-pyridyl ethanol, 5-ethyl-2-2-hydroxyethyl pyridine, 5-ethyl-2-hydroxyethylpyridine, 2-5-ethyl-2-pyridinyl-1-ethanol, 2-pyridineethanol, 5-ethyl, 2-hydroxyethyl-5-ethylpyridine, 5-ethylpyridine-2-ethanol, unii-o6f6rcv39p
PubChem CID 78894
IUPAC Name 2-(5-ethylpyridin-2-yl)ethanol
SMILES CCC1=CN=C(C=C1)CCO

Specifications

Melting Point 44°C
Color White-Yellow
Quantity 5 g
Formula Weight 151.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Ethyl-2-pyridineethanol
EINECSNumber (5)-6671
RTECSNumber UT3150000
TSCA No