missing translation for 'onlineSavingsMsg'
Learn More

5-Ethyl-2-furaldehyde, 98%

Catalog No. AAL0665903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0665903 1 g
AAL0665906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0665903 Supplier Thermo Scientific Chemicals Supplier No. L0665903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 23074-10-4 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23074-10-4
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00030605
InChI Key XADGZBXFWQHBDB-UHFFFAOYSA-N
PubChem CID 89989
IUPAC Name 5-ethylfuran-2-carbaldehyde
SMILES CCC1=CC=C(O1)C=O

Specifications

Density 1.055
Boiling Point 79°C to 81°C (12 mmHg)
Flash Point 65°C (149°F)
Refractive Index 1.522
Quantity 1 g
Beilstein 1982
Sensitivity Air Sensitive
Synonym 5-ethyl-2-furaldehyde, 5-ethylfurfural, 2-furancarboxaldehyde, 5-ethyl, 5-ethyl-furan-2-carbaldehyde, 2-furancarboxaldehyde,5-ethyl, 5-ethylfurfuraldehyde, acmc-1cc1a, 5-ethyl-2-furancarboxaldehyde, 5-ethylfuran-2-carboxaldehyde
Formula Weight 124.14
Percent Purity 98%
Chemical Name or Material 5-Ethyl-2-furaldehyde
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P233-P240-P243-P280-P374-P501c
EINECSNumber 245-411-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only