missing translation for 'onlineSavingsMsg'
Learn More

5-Ethyl-2-(2-hydroxyethyl)pyridine, 98%

Catalog No. p-7047587
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
AAL1948206 5 g
AAL1948214 25 g
2 options

Catalog No. AAL1948206

Supplier: Thermo Scientific Chemicals L1948206

Only null left

CAS: 5223-06-3 | C9H13NO | 151.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5223-06-3
MDL Number MFCD00129040

Specifications

Melting Point 39°C to 44°C
Molecular Formula C6H6N2O2
Quantity 5 g
Beilstein 117340
Synonym 5-pyrimidineacetic acid, 2-pyrimidin-5-yl acetic acid, 5-pyrimidine acetic acid, pyrimidin-5-ylacetic acid, 5-pyrimidineacetate, pubchem13279, pyrimidine-5-acetic acid, pyrimidin-5-yl-acetic acid, pyrimidin-5-yl acetic acid, 5-pyrimidineacetic acid 7ci,8ci,9ci
InChI Key HEEUMCHKVXTQIP-UHFFFAOYSA-N
SMILES C1=C(C=NC=N1)CC(=O)O
IUPAC Name 2-pyrimidin-5-ylacetic acid
Molecular Weight (g/mol) 138.13
PubChem CID 21471675
Formula Weight 151.21
Percent Purity 98%
Chemical Name or Material 5-Ethyl-2-(2-hydroxyethyl)pyridine
Show More Show Less
missing translation for 'healthHazard1' H312-H318-H335
missing translation for 'healthHazard2' GHS H Statement
H311-H318
Toxic in contact with skin.
Causes serious eye damage.
missing translation for 'healthHazard3' P261-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P363-P501c
EINECSNumber 226-024-3
RTECSNumber UT3150000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only