missing translation for 'onlineSavingsMsg'
Learn More

5-Ethyl-2-(2-hydroxyethyl)pyridine, 98%

Catalog No. AAL1948214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1948214 25 g
AAL1948206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1948214 Supplier Thermo Scientific Chemicals Supplier No. L1948214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5223-06-3 | C9H13NO | 151.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5223-06-3
MDL Number MFCD00129040

Specifications

Melting Point 39°C to 44°C
Molecular Formula C9H13NO
Quantity 25 g
Beilstein 117340
Synonym 5-ethyl-2-pyridineethanol, 2-5-ethylpyridin-2-yl ethanol, 2-5-ethyl-2-pyridyl ethanol, 5-ethyl-2-2-hydroxyethyl pyridine, 5-ethyl-2-hydroxyethylpyridine, 2-5-ethyl-2-pyridinyl-1-ethanol, 2-pyridineethanol, 5-ethyl, 2-hydroxyethyl-5-ethylpyridine, 5-ethylpyridine-2-ethanol, unii-o6f6rcv39p
InChI Key OUJMXIPHUCDRAS-UHFFFAOYSA-N
SMILES CCC1=CN=C(C=C1)CCO
IUPAC Name 2-(5-ethylpyridin-2-yl)ethanol
Molecular Weight (g/mol) 151.21
PubChem CID 78894
Formula Weight 151.21
Percent Purity 98%
Chemical Name or Material 5-Ethyl-2-(2-hydroxyethyl)pyridine
Show More Show Less
Hazard Category H312-H318-H335
Hazard Statement GHS H Statement
H311-H318
Toxic in contact with skin.
Causes serious eye damage.
Precautionary Statement P261-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P363-P501c
EINECSNumber 226-024-3
RTECSNumber UT3150000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only