missing translation for 'onlineSavingsMsg'
Learn More

5-Ethoxy-2-fluorophenylboronic Acid (contains varying amounts of Anhydride) 98.0+%, TCI America™

Catalog No. p-7129405
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12351G 1 g
E12355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12351G Supplier TCI America Supplier No. E12351G
Only null left

Chemical Identifiers

CAS 900174-60-9
Molecular Formula C8H10BFO3
Molecular Weight (g/mol) 183.973
MDL Number MFCD05664321
InChI Key ISJGNUFTQTZLEZ-UHFFFAOYSA-N
Synonym 5-Ethoxy-2-fluorobenzeneboronic Acid
PubChem CID 16217468
IUPAC Name (5-ethoxy-2-fluorophenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)OCC)F)(O)O

Specifications

Melting Point 97°C
Color White
Quantity 1 g
Formula Weight 183.97
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Ethoxy-2-fluorophenylboronic Acid (contains varying amounts of Anhydride)