missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Ethoxy-2-fluorophenylboronic Acid (contains varying amounts of Anhydride) 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 97°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 183.97 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Ethoxy-2-fluorophenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
| TSCA | No |