missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Dimethylamino-2-(2-pyridylazo)phenol 98.0+%, TCI America™
$173.37 - $959.98
Chemical Identifiers
| CAS | 50783-80-7 |
|---|---|
| Molecular Formula | C13H14N4O |
| Molecular Weight (g/mol) | 242.282 |
| MDL Number | MFCD00059778 |
| InChI Key | HHPYHNOAMVRXBT-UHFFFAOYSA-N |
| Synonym | 2-(2-Pyridylazo)-5-dimethylaminophenol |
| PubChem CID | 54258744 |
| IUPAC Name | 3-(dimethylamino)-6-(pyridin-2-ylhydrazinylidene)cyclohexa-2,4-dien-1-one |
| SMILES | CN(C)C1=CC(=O)C(=NNC2=CC=CC=N2)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P0910100MG
|
TCI America
P0910100MG |
100 mg |
Each for $173.37
|
|
|||||
|
P09101G
|
TCI America
P09101G |
1 g |
Each for $959.98
|
|
|||||
Chemical Identifiers
| 50783-80-7 | |
| 242.282 | |
| HHPYHNOAMVRXBT-UHFFFAOYSA-N | |
| 54258744 | |
| CN(C)C1=CC(=O)C(=NNC2=CC=CC=N2)C=C1 |
| C13H14N4O | |
| MFCD00059778 | |
| 2-(2-Pyridylazo)-5-dimethylaminophenol | |
| 3-(dimethylamino)-6-(pyridin-2-ylhydrazinylidene)cyclohexa-2,4-dien-1-one |
Specifications
| 50783-80-7 | |
| Purple-Red | |
| MFCD00059778 | |
| 2-(2-Pyridylazo)-5-dimethylaminophenol | |
| CN(C)C1=CC(=O)C(=NNC2=CC=CC=N2)C=C1 | |
| 242.282 | |
| 242.28 | |
| Crystalline Powder |
| 175°C | |
| C13H14N4O | |
| 100 mg | |
| HHPYHNOAMVRXBT-UHFFFAOYSA-N | |
| 3-(dimethylamino)-6-(pyridin-2-ylhydrazinylidene)cyclohexa-2,4-dien-1-one | |
| 54258744 | |
| ≥98.0% (HPLC) | |
| 5-Dimethylamino-2-(2-pyridylazo)phenol [for Determination of Zinc in Serum] |
Safety and Handling
TSCA : No