missing translation for 'onlineSavingsMsg'
Learn More

5-Dimethylamino-2-(2-pyridylazo)phenol 98.0+%, TCI America™

Catalog No. P0910100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0910100MG 100 mg
P09101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0910100MG Supplier TCI America Supplier No. P0910100MG
Only null left

Chemical Identifiers

CAS 50783-80-7
Molecular Formula C13H14N4O
Molecular Weight (g/mol) 242.282
MDL Number MFCD00059778
InChI Key HHPYHNOAMVRXBT-UHFFFAOYSA-N
Synonym 2-(2-Pyridylazo)-5-dimethylaminophenol
PubChem CID 54258744
IUPAC Name 3-(dimethylamino)-6-(pyridin-2-ylhydrazinylidene)cyclohexa-2,4-dien-1-one
SMILES CN(C)C1=CC(=O)C(=NNC2=CC=CC=N2)C=C1

Specifications

Melting Point 175°C
Color Purple-Red
Quantity 100 mg
Formula Weight 242.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Dimethylamino-2-(2-pyridylazo)phenol [for Determination of Zinc in Serum]