missing translation for 'onlineSavingsMsg'
Learn More

5-Diazo-2,2-dimethyl-1,3-dioxane-4,6-dione 98.0+%, TCI America™

Catalog No. d17185g
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
D17185G 5 g
1 options

Catalog No. D17185G

Supplier: TCI America D17185G

Only null left

Chemical Identifiers

CAS 7270-63-5
Molecular Formula C6H6N2O4
Molecular Weight (g/mol) 170.124
MDL Number MFCD00059899
InChI Key YRCVEMYVKSXPLM-UHFFFAOYSA-N
Synonym 5-Diazomeldrum′Cs Acid, Diazomalonic Acid Cyclic Isopropylidine Ester, cycl-Isopropylidene Diazomalonate
PubChem CID 536077
IUPAC Name 5-diazonio-2,2-dimethyl-6-oxo-1,3-dioxin-4-olate
SMILES CC1(OC(=C(C(=O)O1)[N+]#N)[O-])C

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 5 g
UN Number 1325
Formula Weight 170.12
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Diazo-2,2-dimethyl-1,3-dioxane-4,6-dione
TSCA No
Recommended Storage Refrigerator