missing translation for 'onlineSavingsMsg'
Learn More

5-(Di-tert-butylphosphino)-1',3',5'-triphenyl-1'H-1,4-bipyrazole, 97%

Catalog No. AC435160010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435160010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC435160010 Supplier Thermo Scientific Chemicals Supplier No. 435160010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 894086-00-1 | C32H35N4P | 506.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 894086-00-1
Molecular Formula C32H35N4P
Molecular Weight (g/mol) 506.62
InChI Key PTXJGGGNGMPMBG-UHFFFAOYSA-N
Synonym bippyphos, t-bu-bippyphos, 5-di-tert-butylphosphino-1',3',5'-triphenyl-1'h-1,4'-bipyrazole, 5-di-tert-butylphosphino-1', 3', 5'-triphenyl-1'h-1,4' bipyrazole, 5-di-tert-butylphosphino-1',3',5'-triphenyl-1'h-1,4' bipyrazole, 5-di-t-butylphosphino-1',3',5'-triphenyl-1,4'-bi-1h-pyrazole, 5-bis tert-butyl phosphino-1',3',5'-triphenyl-1,4'-bi-1h-pyrazole, 5-di-tert-butylphosphanyl-1',3',5'-triphenyl-1,4'-bipyrazole, 1',3',5'-triphenyl-5-ditert-butylphosphino-1,4'-bi 1h-pyrazole, 5-di-tert-butyl-phosphanyl-1',3',5'-triphenyl-1'h-1,4' bipyrazolyl
PubChem CID 11547931
IUPAC Name ditert-butyl-[2-(1,3,5-triphenylpyrazol-4-yl)pyrazol-3-yl]phosphane
SMILES CC(C)(C)P(C1=CC=NN1C2=C(N(N=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5)C(C)(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 169.0°C to 173.0°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 506.62
Percent Purity 97%
Physical Form Crystalline Powder or Solid
Chemical Name or Material 5-(Di-tert-Butylphosphino)-1',3',5'-triphenyl-1'H-1,4-bipyrazole
Show More Show Less

RUO – Research Use Only