missing translation for 'onlineSavingsMsg'
Learn More

5-Decyne 98.0+%, TCI America™

Catalog No. D11905ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11905ML 5 mL
D119025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D11905ML Supplier TCI America Supplier No. D11905ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1942-46-7
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00027277
InChI Key JWBQJUFCNOLNNC-UHFFFAOYSA-N
Synonym 5-decyne, dibutylacetylene, 1,2-dibutylacetylene, acmc-209ey1, 5-decyne 8ci 9ci, 3,3?-diheptyloxacarbocyanine iodide
PubChem CID 16030
IUPAC Name dec-5-yne
SMILES CCCCC#CCCCC

Specifications

Color Yellow
Boiling Point 177°C
Quantity 5 mL
UN Number 3295
Formula Weight 138.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Decyne