missing translation for 'onlineSavingsMsg'
Learn More

5-Decyn-1-ol, 97%

Catalog No. p-7035677
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5340006 5 g
AAH5340014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5340006 Supplier Thermo Scientific Chemicals Supplier No. H5340006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 68274-97-5 | C10H18O | 154.25 g/mol

5-Decyn-1-ol is used in the synthesis of multi-substituted cyclopropanes. It involved in manufacture of fine chemicals, having applications in electroplating and perfumery industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Decyn-1-ol is used in the synthesis of multi-substituted cyclopropanes. It involved in manufacture of fine chemicals, having applications in electroplating and perfumery industry.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 68274-97-5
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00464353
InChI Key RNTNGQCWAKHVKO-UHFFFAOYSA-N
Synonym 5-decyn-1-ol, 5-decyn-1-ol,
PubChem CID 109957
IUPAC Name dec-5-yn-1-ol
SMILES CCCCC#CCCCCO

Specifications

Density 0.861
Boiling Point 238°C
Refractive Index 1.461
Linear Formula CH3(CH2)3C≡C(CH2)4OH
Quantity 5 g
Formula Weight 154.25
Percent Purity 97%
Chemical Name or Material 5-Decyn-1-ol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 269-529-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only