missing translation for 'onlineSavingsMsg'
Learn More

5-Cyclopropyl-1,2,4-oxadiazol-3-amine, 97%, Thermo Scientific™

Catalog No. p-7077336
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450030010 1g
AC450030050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC450030010 Supplier Thermo Scientific Chemicals Supplier No. 450030010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 868696-42-8
Molecular Formula C5H7N3O
Molecular Weight (g/mol) 125.13
InChI Key OHSZRALIDGBNES-UHFFFAOYSA-N
Synonym 1,2,4-oxadiazol-3-amine, 5-cyclopropyl
PubChem CID 51072255
IUPAC Name 5-cyclopropyl-1,2,4-oxadiazol-3-amine
SMILES C1CC1C2=NC(=NO2)N

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1g
Formula Weight 125.13
Percent Purity 97%
Chemical Name or Material 5-Cyclopropyl-1,2,4-oxadiazol-3-amine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only