missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Cyclopropyl-1,2,4-oxadiazol-3-amine, 97%, Thermo Scientific™
Specifications
Specifications
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (HPLC) |
| Packaging | Glass Bottle |
| Quantity | 1g |
| Formula Weight | 125.13 |
| Percent Purity | 97% |
| Chemical Name or Material | 5-Cyclopropyl-1,2,4-oxadiazol-3-amine |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. |
RUO – Research Use Only