missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Cyclohexadecen-1-one 97.0+%, TCI America™
$55.92 - $171.49
Chemical Identifiers
| CAS | 37609-25-9 |
|---|---|
| Molecular Formula | C16H28O |
| Molecular Weight (g/mol) | 236.40 |
| MDL Number | MFCD00059511 |
| InChI Key | ABRIMXGLNHCLIP-VURMDHGXSA-N |
| PubChem CID | 6506221 |
| IUPAC Name | (5Z)-cyclohexadec-5-en-1-one |
| SMILES | O=C1CCCCCCCCCC\C=C/CCC1 |
Chemical Identifiers
| 37609-25-9 | |
| 236.40 | |
| ABRIMXGLNHCLIP-VURMDHGXSA-N | |
| (5Z)-cyclohexadec-5-en-1-one |
| C16H28O | |
| MFCD00059511 | |
| 6506221 | |
| O=C1CCCCCCCCCC\C=C/CCC1 |
Specifications
| 37609-25-9 | |
| 120°C | |
| MFCD00059511 | |
| ABRIMXGLNHCLIP-VURMDHGXSA-N | |
| (5Z)-cyclohexadec-5-en-1-one | |
| 6506221 | |
| ≥97.0% (GC) | |
| 5-Cyclohexadecen-1-one |
| White | |
| C16H28O | |
| 5 g | |
| O=C1CCCCCCCCCC\C=C/CCC1 | |
| 236.40 | |
| 236.40 | |
| Crystalline Lumps |
Safety and Handling
TSCA : Yes