missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Cyclohexadecen-1-one 97.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Boiling Point | 120°C |
| Quantity | 5 g |
| Formula Weight | 236.40 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 5-Cyclohexadecen-1-one |
Safety and Handling
| TSCA | Yes |