missing translation for 'onlineSavingsMsg'
Learn More

5-Cyanothiophene-2-boronic acid, 98%, Thermo Scientific™

Chemical Identifiers

CAS 305832-67-1
Molecular Formula C5H4BNO2S
Molecular Weight (g/mol) 152.97
MDL Number MFCD02094029
InChI Key ZEOMEPSYIIQIND-UHFFFAOYSA-N
Synonym 5-cyanothiophene-2-boronic acid, 5-cyanothiophen-2-yl boronic acid, 5-cyanothiophen-2-yl-2-boronic acid, 5-boronothiophene-2-carbonitrile, 5-cyano-2-thienyl boronic acid, 5-dihydroxyboranyl thiophene-2-carbonitrile, 5-cyano-2-thienylboronic acid, boronic acid, 5-cyano-2-thienyl, 5-cyanothiophen-2-boronic acid, acmc-1ctts
PubChem CID 4198743
IUPAC Name (5-cyanothiophen-2-yl)boronic acid
SMILES B(C1=CC=C(S1)C#N)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC391270010
SDS View Documents
Thermo Scientific Chemicals
391270010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC391270050
SDS View Documents
Thermo Scientific Chemicals
391270050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

305832-67-1
152.97
ZEOMEPSYIIQIND-UHFFFAOYSA-N
4198743
B(C1=CC=C(S1)C#N)(O)O
C5H4BNO2S
MFCD02094029
5-cyanothiophene-2-boronic acid, 5-cyanothiophen-2-yl boronic acid, 5-cyanothiophen-2-yl-2-boronic acid, 5-boronothiophene-2-carbonitrile, 5-cyano-2-thienyl boronic acid, 5-dihydroxyboranyl thiophene-2-carbonitrile, 5-cyano-2-thienylboronic acid, boronic acid, 5-cyano-2-thienyl, 5-cyanothiophen-2-boronic acid, acmc-1ctts
(5-cyanothiophen-2-yl)boronic acid

Specifications

305832-67-1
100.0
97.5% min. (HPLC)
C5H4BNO2S
1g
Solubility in water: insoluble. Other solubilities: soluble in ethanol and thf
B(C1=CC=C(S1)C#N)(O)O
152.97
152.97
5-Cyanothiophene-2-boronic acid
97.5
Authentic
Glass bottle
MFCD02094029
5-cyanothiophene-2-boronic acid, 5-cyanothiophen-2-yl boronic acid, 5-cyanothiophen-2-yl-2-boronic acid, 5-boronothiophene-2-carbonitrile, 5-cyano-2-thienyl boronic acid, 5-dihydroxyboranyl thiophene-2-carbonitrile, 5-cyano-2-thienylboronic acid, boronic acid, 5-cyano-2-thienyl, 5-cyanothiophen-2-boronic acid, acmc-1ctts
ZEOMEPSYIIQIND-UHFFFAOYSA-N
(5-cyanothiophen-2-yl)boronic acid
4198743
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only