missing translation for 'onlineSavingsMsg'
Learn More

5-Cyano-2-methylpyridine 98.0+%, TCI America™

Catalog No. p-7125801
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30501G 1 g
C30505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30501G Supplier TCI America Supplier No. C30501G
Only null left

Chemical Identifiers

CAS 3222-48-8
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00038042
InChI Key PBLOYQAQGYUPCM-UHFFFAOYSA-N
Synonym 5-cyano-2-picoline, 5-cyano-2-methylpyridine, 6-methylnicotinonitrile, 6-methyl-3-pyridinecarbonitrile, 3-cyano-6-methylpyridine, 3-pyridinecarbonitrile, 6-methyl, pubchem6695, 2-methyl-5-cyanopyridine, 6-methylnicotinonitrile #, acmc-209z1l
PubChem CID 281995
IUPAC Name 6-methylpyridine-3-carbonitrile
SMILES CC1=NC=C(C=C1)C#N

Specifications

Melting Point 84°C
Color Yellow
Boiling Point 217°C
Quantity 1 g
UN Number 3439
Formula Weight 118.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Cyano-2-methylpyridine