missing translation for 'onlineSavingsMsg'
Learn More

5-Cyano-2-methylpyridine, 99%

Catalog No. AAL0987503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0987503 1 g
AAL0987506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0987503 Supplier Thermo Scientific Chemicals Supplier No. L0987503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3222-48-8 | C7H6N2 | 118.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3222-48-8
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00038042
InChI Key PBLOYQAQGYUPCM-UHFFFAOYSA-N
Synonym 5-cyano-2-picoline, 5-cyano-2-methylpyridine, 6-methylnicotinonitrile, 6-methyl-3-pyridinecarbonitrile, 3-cyano-6-methylpyridine, 3-pyridinecarbonitrile, 6-methyl, pubchem6695, 2-methyl-5-cyanopyridine, 6-methylnicotinonitrile #, acmc-209z1l
PubChem CID 281995
IUPAC Name 6-methylpyridine-3-carbonitrile
SMILES CC1=NC=C(C=C1)C#N

Specifications

Melting Point 83°C to 87°C
Boiling Point 216°C to 217°C
Quantity 1 g
Beilstein 111235
Formula Weight 118.14
Percent Purity 99%
Chemical Name or Material 5-Cyano-2-methylpyridine
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only