missing translation for 'onlineSavingsMsg'
Learn More

5-Cyano-2-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7193750
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C32271G Supplier TCI America Supplier No. C32271G

Chemical Identifiers

CAS 468718-30-1
Molecular Formula C7H5BFNO2
Molecular Weight (g/mol) 164.93
MDL Number MFCD03095147
InChI Key ITCSMTHUTFTXLE-UHFFFAOYSA-N
PubChem CID 2783361
IUPAC Name (5-cyano-2-fluorophenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)C#N)F)(O)O

Specifications

Color White
Quantity 1 g
Synonym 5-cyano-2-fluorobenzeneboronic acid, 5-cyano-2-fluorophenyl boronic acid, 3-borono-4-fluorobenzonitrile, boronic acid, 5-cyano-2-fluorophenyl, 5-cyano-2-fluoro-phenyl boronic acid, boronic acid, b-5-cyano-2-fluorophenyl, 5-cyano-2-fluorobenzeneboronicacid, pubchem1807, pubchem7788, acmc-1apk5
Formula Weight 164.93
Physical Form Crystalline Powder
Chemical Name or Material 5-Cyano-2-fluorophenylboronic Acid (contains varying amounts of Anhydride)