missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorovaleryl Chloride 98.0+%, TCI America™

Catalog No. C17895G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17895G 5 g
C178925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17895G Supplier TCI America Supplier No. C17895G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1575-61-7
Molecular Formula C5H8Cl2O
Molecular Weight (g/mol) 155.02
MDL Number MFCD00000758
InChI Key SVNNWKWHLOJLOK-UHFFFAOYSA-N
Synonym 5-chlorovaleryl chloride, valeryl chloride, 5-chloro, pentanoyl chloride, 5-chloro, 5-chlorovalerylchloride, 5-chlorovaleroyl chloride, pentanoylchloride, 5-chloro, chlorovaleryl chloride, pubchem10919, 5-chloro-pentanoylchloride, acmc-1cuj1
PubChem CID 74089
IUPAC Name 5-chloropentanoyl chloride
SMILES ClCCCCC(Cl)=O

Specifications

Color Yellow
Quantity 5 g
UN Number 3265
Formula Weight 155.02
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Chlorovaleryl Chloride
RTECSNumber YV9108000
TSCA No