missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorovaleroyl chloride, 98%

Catalog No. AAH5623718
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5623718 50 g
AAH5623722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5623718 Supplier Thermo Scientific Chemicals Supplier No. H5623718

CAS: 1575-61-7 | C5H8Cl2O | 155.02 g/mol

5-Chlorovaleroyl chloride (5-CVC) has been used in the development and validation of a GC-FID method for the analysis of low level impurities present in 5-CVC. It has been used in the synthesis of anion-exchange resins by acylation of polystyrene/divinylbenzene and subsequent amination with trimethylamine. It is commonly used as alkylating agent in the synthesis of pharmaceutical intermediates and other specialty chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chlorovaleroyl chloride (5-CVC) has been used in the development and validation of a GC-FID method for the analysis of low level impurities present in 5-CVC. It has been used in the synthesis of anion-exchange resins by acylation of polystyrene/divinylbenzene and subsequent amination with trimethylamine. It is commonly used as alkylating agent in the synthesis of pharmaceutical intermediates and other specialty chemicals.

Solubility
Reacts with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, bases, alcohols, amines.

Chemical Identifiers

CAS 1575-61-7
Molecular Formula C5H8Cl2O
Molecular Weight (g/mol) 155.02
MDL Number MFCD00000758
InChI Key SVNNWKWHLOJLOK-UHFFFAOYSA-N
Synonym 5-chlorovaleryl chloride, valeryl chloride, 5-chloro, pentanoyl chloride, 5-chloro, 5-chlorovalerylchloride, 5-chlorovaleroyl chloride, pentanoylchloride, 5-chloro, chlorovaleryl chloride, pubchem10919, 5-chloro-pentanoylchloride, acmc-1cuj1
PubChem CID 74089
IUPAC Name 5-chloropentanoyl chloride
SMILES ClCCCCC(Cl)=O

Specifications

Density 1.2
Boiling Point 38°C to 39°C
Flash Point 91°C (196°F)
Refractive Index 1.464
Linear Formula Cl(CH2)4COCl
Quantity 50 g
UN Number UN3265
Beilstein 1745182
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 155.02
Percent Purity 98%
Chemical Name or Material 5-Chlorovaleroyl chloride
Show More Show Less
Hazard Category H227-H302-H314
Hazard Statement GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 216-403-1
RTECSNumber YV9108000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only