missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorovaleronitrile 96.0+%, TCI America™

Catalog No. p-7124970
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C142125G 25 g
C1421250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C142125G Supplier TCI America Supplier No. C142125G
Only null left

Chemical Identifiers

CAS 6280-87-1
Molecular Formula C5H8ClN
Molecular Weight (g/mol) 117.58
MDL Number MFCD00001977
InChI Key JSAWFGSXRPCFSW-UHFFFAOYSA-N
Synonym 5-chlorovaleronitrile, pentanenitrile, 5-chloro, 5-chloro-n-valeronitrile, valeronitrile, 5-chloro, 5-chloro valeronitrile, .delta.-chlorovaleronitrile, snxhfhaiuzjp@, 5-chlorovaieronitrile, 4-chlorobutyl cyanide, delta-chlorovaleronitrile
PubChem CID 80474
IUPAC Name 5-chloropentanenitrile
SMILES ClCCCCC#N

Specifications

Color Yellow
Boiling Point 118°C
Quantity 25 g
UN Number 3276
Formula Weight 117.58
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Chlorovaleronitrile