missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorovaleronitrile, 97%, Thermo Scientific™

$214.04 - $214.04

Chemical Identifiers

CAS 6280-87-1
Molecular Formula C5H8ClN
Molecular Weight (g/mol) 117.58
MDL Number MFCD00001977
InChI Key JSAWFGSXRPCFSW-UHFFFAOYSA-N
Synonym 5-chlorovaleronitrile, pentanenitrile, 5-chloro, 5-chloro-n-valeronitrile, valeronitrile, 5-chloro, 5-chloro valeronitrile, .delta.-chlorovaleronitrile, snxhfhaiuzjp@, 5-chlorovaieronitrile, 4-chlorobutyl cyanide, delta-chlorovaleronitrile
PubChem CID 80474
IUPAC Name 5-chloropentanenitrile
SMILES ClCCCCC#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC110140250
SDS View Documents
Thermo Scientific Chemicals
110140250
25 mL Glass bottle N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6280-87-1
117.58
JSAWFGSXRPCFSW-UHFFFAOYSA-N
80474
ClCCCCC#N
C5H8ClN
MFCD00001977
5-chlorovaleronitrile, pentanenitrile, 5-chloro, 5-chloro-n-valeronitrile, valeronitrile, 5-chloro, 5-chloro valeronitrile, .delta.-chlorovaleronitrile, snxhfhaiuzjp@, 5-chlorovaieronitrile, 4-chlorobutyl cyanide, delta-chlorovaleronitrile
5-chloropentanenitrile

Specifications

6280-87-1
100.0
92°C to 93°C (10.0 mmHg)
Authentic
Glass bottle
1.4452 to 1.4472
MFCD00001977
1.04
JSAWFGSXRPCFSW-UHFFFAOYSA-N
5-chloropentanenitrile
80474
97%
5-Chlorovaleronitrile, 97%
96.0
1.0400g/mL
98°C
96% min. (GC)
C5H8ClN
Cl(CH2)4CN
25 mL
5-chlorovaleronitrile, pentanenitrile, 5-chloro, 5-chloro-n-valeronitrile, valeronitrile, 5-chloro, 5-chloro valeronitrile, .delta.-chlorovaleronitrile, snxhfhaiuzjp@, 5-chlorovaieronitrile, 4-chlorobutyl cyanide, delta-chlorovaleronitrile
ClCCCCC#N
117.58
117.58
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 228-481-4

Recommended Storage : Product darkens on storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only