missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorothiophene-2-sulfonyl chloride, 97%

CAS: 2766-74-7 | C4H2Cl2O2S2 | 217.08 g/mol

Supplier:  Thermo Scientific Chemicals A1134806

 View more versions of this product

Catalog No. AAA1134806


Only null left
Explore available promotions

Description

Description

5-Chlorothiophene-2-sulfonyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs. It is used in the preparation of Notch-1-sparing γ-secretase inhibitors for the treatment of Alzheimer's disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Not miscible in water.

Notes
Moisture Sensitive. Store at 4°C. Incomaptible with oxidizing agents and bases.
Specifications

Specifications

5-Chlorothiophene-2-sulfonyl chloride
1.623
>110°C (230°F)
1.586
5 g
130710
5-chlorothiophene-2-sulfonylchloride, 5-chloro-2-thiophenesulfonyl chloride, 5-chlorothiophene-2-sulphonyl chloride, 5-chlorothiophenesulphonyl chloride, 5-chloro-2-thienylsulfonyl chloride, 5-chloro-thiophene-2-sulfonyl chloride, 5-chlorothiophenesulfonyl chloride, 5-chlorothiophen-2-sulfonyl chloride, 5-chlorothiophene-2-sulphonylchloride, 2-thiophenesulfonyl chloride, 5-chloro
SORSTNOXGOXWAO-UHFFFAOYSA-N
5-chlorothiophene-2-sulfonyl chloride
2733925
97%
2766-74-7
117°C to 120°C (14 mmHg)
C4H2Cl2O2S2
MFCD00051667
UN3265
Moisture sensitive
Not miscible in water.
ClC1=CC=C(S1)S(Cl)(=O)=O
217.08
217.09

Chemical Identifiers

2766-74-7
217.08
SORSTNOXGOXWAO-UHFFFAOYSA-N
2733925
ClC1=CC=C(S1)S(Cl)(=O)=O
C4H2Cl2O2S2
MFCD00051667
5-chlorothiophene-2-sulfonylchloride, 5-chloro-2-thiophenesulfonyl chloride, 5-chlorothiophene-2-sulphonyl chloride, 5-chlorothiophenesulphonyl chloride, 5-chloro-2-thienylsulfonyl chloride, 5-chloro-thiophene-2-sulfonyl chloride, 5-chlorothiophenesulfonyl chloride, 5-chlorothiophen-2-sulfonyl chloride, 5-chlorothiophene-2-sulphonylchloride, 2-thiophenesulfonyl chloride, 5-chloro
5-chlorothiophene-2-sulfonyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 700-248-6

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only