missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorothiophene-2-carboxylic acid, 98%

Catalog No. AAA1520906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1520906 5 g
AAA1520914 25 g
AAA1520922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1520906 Supplier Thermo Scientific Chemicals Supplier No. A1520906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 24065-33-6 | C5H3ClO2S | 162.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24065-33-6
Molecular Formula C5H3ClO2S
Molecular Weight (g/mol) 162.59
MDL Number MFCD00041426
InChI Key QZLSBOVWPHXCLT-UHFFFAOYSA-N
Synonym 5-chloro-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-chloro, 2-chloro-5-carboxythiophene, 5-chloro-2-thiophene carboxylic acid, pubchem7380, maybridge3_003717, acmc-209g8i, rivaroxaban related compound, 5-18-06-00177 beilstein handbook reference, ksc204m2j
PubChem CID 95048
IUPAC Name 5-chlorothiophene-2-carboxylic acid
SMILES OC(=O)C1=CC=C(Cl)S1

Specifications

Melting Point 149°C to 152°C
Quantity 5 g
Beilstein 118361
Formula Weight 162.6
Percent Purity 98%
Chemical Name or Material 5-Chlorothiophene-2-carboxylic acid
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
EINECSNumber 000-000-0
RTECSNumber XM8400000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only