missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorothiophene-2-carboxaldehyde 95.0+%, TCI America™

Catalog No. C17221G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17221G 1 g
C17225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17221G Supplier TCI America Supplier No. C17221G

Chemical Identifiers

CAS 7283-96-7
Molecular Formula C5H3ClOS
Molecular Weight (g/mol) 146.59
MDL Number MFCD00047090
InChI Key VWYFITBWBRVBSW-UHFFFAOYSA-N
Synonym 5-chloro-2-thiophenecarboxaldehyde, 2-chloro-5-thiophenecarboxaldehyde, 5-chlorothiophene-2-carboxaldehyde, 2-thiophenecarboxaldehyde, 5-chloro, 5-chloro-2-formylthiophene, 2-chloro-5-formylthiophene, 5-chlorothiophene-2-aldehyde, 5-chloro-2-thiophenecarbaldehyde, 5-chloro-2-thiophene-carboxaldehyde, pubchem7742
PubChem CID 81700
IUPAC Name 5-chlorothiophene-2-carbaldehyde
SMILES ClC1=CC=C(S1)C=O

Specifications

Color Brown-Yellow
Quantity 1 g
Formula Weight 146.59
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Chlorothiophene-2-carboxaldehyde