missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorothiophene-2-carbonyl Chloride 98.0+%, TCI America™

Catalog No. p-7125689
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
C28645G 5 g
C286425G 25 g
2 options

Catalog No. C28645G

Supplier: TCI America C28645G

Only null left

Chemical Identifiers

CAS 42518-98-9
Molecular Formula C5H2Cl2OS
Molecular Weight (g/mol) 181.03
MDL Number MFCD00043886
InChI Key BMPDCQVRKDNUAP-UHFFFAOYSA-N
Synonym 5-chlorothiophene-2-carbonylchloride, 5-chorothiophene-2-carbonyl chloride, 5-chloro-2-thiophenecarbonyl chloride, 5-chloro-thiophene-2-carbonyl chloride, 5-chloro-2-thenoyl chloride, 2-thiophenecarbonyl chloride, 5-chloro, pubchem16132, ksc235i7t, 5-chlorothiophenecarbonyl chloride, 5-chloro-2-thenoylchloride
PubChem CID 589223
IUPAC Name 5-chlorothiophene-2-carbonyl chloride
SMILES ClC(=O)C1=CC=C(Cl)S1

Specifications

Melting Point 4°C
Color Yellow
Boiling Point 225°C
Quantity 5 g
UN Number 3265
Formula Weight 181.03
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Chlorothiophene-2-carbonyl Chloride