missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorothiophene-2-carbonyl chloride, 98%

Catalog No. AAH6147414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6147414 25 g
AAH6147406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6147414 Supplier Thermo Scientific Chemicals Supplier No. H6147414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 42518-98-9 | C5H2Cl2OS | 181.03 g/mol

5-Chlorothiophene-2-carbonyl chloride used as a intermediate for rivaroxaban.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chlorothiophene-2-carbonyl chloride used as a intermediate for rivaroxaban.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store in cool, dry place in tightly closed container. Store away form water/moisture.

Chemical Identifiers

CAS 42518-98-9
Molecular Formula C5H2Cl2OS
Molecular Weight (g/mol) 181.03
MDL Number MFCD00043886
InChI Key BMPDCQVRKDNUAP-UHFFFAOYSA-N
Synonym 5-chlorothiophene-2-carbonylchloride, 5-chorothiophene-2-carbonyl chloride, 5-chloro-2-thiophenecarbonyl chloride, 5-chloro-thiophene-2-carbonyl chloride, 5-chloro-2-thenoyl chloride, 2-thiophenecarbonyl chloride, 5-chloro, pubchem16132, ksc235i7t, 5-chlorothiophenecarbonyl chloride, 5-chloro-2-thenoylchloride
PubChem CID 589223
IUPAC Name 5-chlorothiophene-2-carbonyl chloride
SMILES ClC(=O)C1=CC=C(Cl)S1

Specifications

Melting Point 4°C
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 181.04
Percent Purity 98%
Chemical Name or Material 5-Chlorothiophene-2-carbonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only