missing translation for 'onlineSavingsMsg'
Learn More

5-chlorosulfonyl-2-hydroxybenzoic acid, 97%, Thermo Scientific™

Catalog No. p-2702965
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CD11776DA 1g
CD11776DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CD11776DA Supplier Thermo Scientific Chemicals Supplier No. CD11776DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17243-13-9
Molecular Formula C7H5ClO5S
Molecular Weight (g/mol) 236.62
MDL Number MFCD00052974
InChI Key SJXHSFSHNKFRLN-UHFFFAOYSA-N
Synonym 5-chlorosulfonyl-2-hydroxybenzoic acid, 5-chlorosulfonyl-2-hydroxy-benzoic acid, 5-chlorosulfonyl salicylic acid, benzoic acid, 5-chlorosulfonyl-2-hydroxy, benzoic acid,5-chlorosulfonyl-2-hydroxy, 5-chlorosulphonyl-2-hydroxybenzoic acid, 5-chlorosulfonyl-2-hydroxybenzoicacid, acmc-1cis5, 5-chlorosulphonyl salicylic acid
PubChem CID 2777240
IUPAC Name 5-chlorosulfonyl-2-hydroxybenzoic acid
SMILES OC(=O)C1=C(O)C=CC(=C1)S(Cl)(=O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1g
Formula Weight 236.63
Percent Purity 97%
Chemical Name or Material 5-Chlorosulfonyl-2-hydroxybenzoic acid
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.