missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorosalicylaldehyde 98.0+%, TCI America™

Catalog No. C07385G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07385G 5 g
C073825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07385G Supplier TCI America Supplier No. C07385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 635-93-8
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.57
MDL Number MFCD00003331
InChI Key FUGKCSRLAQKUHG-UHFFFAOYSA-N
Synonym 5-chlorosalicylaldehyde, 2-hydroxy-5-chlorobenzaldehyde, benzaldehyde, 5-chloro-2-hydroxy, 4-chloro-2-formylphenol, salicylaldehyde, 5-chloro, 5-chloro-2-hydroxy-benzaldehyde, unii-4gcc8zkm3o, 5-chloro-salicylaldehyde, 4gcc8zkm3o, 5-chloro-2-hydroxy benzaldehyde
PubChem CID 12481
IUPAC Name 5-chloro-2-hydroxybenzaldehyde
SMILES OC1=CC=C(Cl)C=C1C=O

Specifications

Melting Point 101°C
Color White-Yellow
Quantity 5 g
Formula Weight 156.57
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chlorosalicylaldehyde
EINECSNumber (3)-1192
RTECSNumber VN5450000
TSCA Yes