missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorosalicylaldehyde, 98%

Catalog No. AAA1406906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1406906 5 g
AAA1406914 25 g
AAA1406922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1406906 Supplier Thermo Scientific Chemicals Supplier No. A1406906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 635-93-8 | C7H5ClO2 | 156.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 635-93-8
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.57
MDL Number MFCD00003331
InChI Key FUGKCSRLAQKUHG-UHFFFAOYSA-N
Synonym 5-chlorosalicylaldehyde, 2-hydroxy-5-chlorobenzaldehyde, benzaldehyde, 5-chloro-2-hydroxy, 4-chloro-2-formylphenol, salicylaldehyde, 5-chloro, 5-chloro-2-hydroxy-benzaldehyde, unii-4gcc8zkm3o, 5-chloro-salicylaldehyde, 4gcc8zkm3o, 5-chloro-2-hydroxy benzaldehyde
PubChem CID 12481
IUPAC Name 5-chloro-2-hydroxybenzaldehyde
SMILES OC1=CC=C(Cl)C=C1C=O

Specifications

Melting Point 99°C to 103°C
Quantity 5 g
Beilstein 636632
Sensitivity Air Sensitive
Formula Weight 156.57
Percent Purity 98%
Chemical Name or Material 5-Chlorosalicylaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 211-244-4
RTECSNumber VN5450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only