missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorosalicylaldehyde, 98%

CAS: 635-93-8 | C7H5ClO2 | 156.57 g/mol

Supplier:  Thermo Scientific Chemicals 405030250

 View more versions of this product

Catalog No. AC405030250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Chlorosalicylaldehyde
635-93-8
100.0
White to Yellow
97.5% min. (GC)
C7H5ClO2
25 g
Solubility in water: insoluble
OC1=CC=C(Cl)C=C1C=O
156.57
156.57
Crystalline Powder
98%
97.5
102°C to 103°C
Authentic
Plastic bottle
MFCD00003331
5-chlorosalicylaldehyde, 2-hydroxy-5-chlorobenzaldehyde, benzaldehyde, 5-chloro-2-hydroxy, 4-chloro-2-formylphenol, salicylaldehyde, 5-chloro, 5-chloro-2-hydroxy-benzaldehyde, unii-4gcc8zkm3o, 5-chloro-salicylaldehyde, 4gcc8zkm3o, 5-chloro-2-hydroxy benzaldehyde
FUGKCSRLAQKUHG-UHFFFAOYSA-N
5-chloro-2-hydroxybenzaldehyde
12481
98%

Chemical Identifiers

635-93-8
156.57
FUGKCSRLAQKUHG-UHFFFAOYSA-N
12481
OC1=CC=C(Cl)C=C1C=O
C7H5ClO2
MFCD00003331
5-chlorosalicylaldehyde, 2-hydroxy-5-chlorobenzaldehyde, benzaldehyde, 5-chloro-2-hydroxy, 4-chloro-2-formylphenol, salicylaldehyde, 5-chloro, 5-chloro-2-hydroxy-benzaldehyde, unii-4gcc8zkm3o, 5-chloro-salicylaldehyde, 4gcc8zkm3o, 5-chloro-2-hydroxy benzaldehyde
5-chloro-2-hydroxybenzaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Very toxic to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid releas

GHS Signal Word: Warning

EINECSNumber : 211-244-4

RTECSNumber : VN5450000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only