missing translation for 'onlineSavingsMsg'
Learn More

5-Chloropyrimidine, 95%

Catalog No. AC459660050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459660050 5 g
AC459660010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459660050 Supplier Thermo Scientific Chemicals Supplier No. 459660050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17180-94-8 | C4H3ClN2 | 114.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17180-94-8
Molecular Formula C4H3ClN2
Molecular Weight (g/mol) 114.53
MDL Number MFCD03840717
InChI Key ZLNPDTOTEVIMMY-UHFFFAOYSA-N
Synonym pyrimidine, 5-chloro, 5-chloro-pyrimidine, 5-choloropyrimidine, 5-chloranylpyrimidine, pubchem7041, pyrimidine, 5-chloro-6ci,8ci,9ci
PubChem CID 12600384
IUPAC Name 5-chloropyrimidine
SMILES ClC1=CN=CN=C1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 114.53
Percent Purity 95%
Chemical Name or Material 5-Chloropyrimidine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POI

RUO – Research Use Only