missing translation for 'onlineSavingsMsg'
Learn More

5-Chloropyridine-2-boronic acid, 95%

CAS: 652148-91-9 | C5H5BClNO2 | 157.36 g/mol

$111.22 - $377.78

Chemical Identifiers

CAS 652148-91-9
Molecular Formula C5H5BClNO2
Molecular Weight (g/mol) 157.36
MDL Number MFCD07368862
InChI Key JEGHCYRKSUGHJH-UHFFFAOYSA-N
Synonym 5-chloro-2-pyridineboronic acid, 5-chloropyridine-2-boronic acid, 5-chloropyridin-2-yl boronic acid, 2-borono-5-chloropyridine, 5-chloropyridine-2-boronicacid, 3-chloropyridine-6-boronic acid, 5-chloro-2-pyridyl boronic acid, boronic acid, 5-chloro-2-pyridinyl, pubchem13568, acmc-209npx
PubChem CID 22832086
IUPAC Name (5-chloropyridin-2-yl)boronic acid
SMILES B(C1=NC=C(C=C1)Cl)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH52515MD
View Documents
Thermo Scientific Chemicals
H52515MD
250 mg N/A
Only null left
 
AAH5251503
View Documents
Thermo Scientific Chemicals
H5251503
1 g N/A
Only null left
 
Description

Description

5-Chloropyridine-2-boronic acid was used as reagent in organic synthesis. Also used as phamaceutical Intermediates. Used in suzuki-Miyaura coupling processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloropyridine-2-boronic acid was used as reagent in organic synthesis. Also used as phamaceutical Intermediates. Used in suzuki-Miyaura coupling processes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

652148-91-9
157.36
JEGHCYRKSUGHJH-UHFFFAOYSA-N
22832086
B(C1=NC=C(C=C1)Cl)(O)O
C5H5BClNO2
MFCD07368862
5-chloro-2-pyridineboronic acid, 5-chloropyridine-2-boronic acid, 5-chloropyridin-2-yl boronic acid, 2-borono-5-chloropyridine, 5-chloropyridine-2-boronicacid, 3-chloropyridine-6-boronic acid, 5-chloro-2-pyridyl boronic acid, boronic acid, 5-chloro-2-pyridinyl, pubchem13568, acmc-209npx
(5-chloropyridin-2-yl)boronic acid

Specifications

652148-91-9
MFCD07368862
5-chloro-2-pyridineboronic acid, 5-chloropyridine-2-boronic acid, 5-chloropyridin-2-yl boronic acid, 2-borono-5-chloropyridine, 5-chloropyridine-2-boronicacid, 3-chloropyridine-6-boronic acid, 5-chloro-2-pyridyl boronic acid, boronic acid, 5-chloro-2-pyridinyl, pubchem13568, acmc-209npx
B(C1=NC=C(C=C1)Cl)(O)O
157.36
157.37
5-Chloropyridine-2-boronic acid
C5H5BClNO2
250 mg
JEGHCYRKSUGHJH-UHFFFAOYSA-N
(5-chloropyridin-2-yl)boronic acid
22832086
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only