missing translation for 'onlineSavingsMsg'
Learn More

5-Chloropyridine-2-boronic acid, 95%

Catalog No. AAH52515MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52515MD 250 mg
AAH5251503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52515MD Supplier Thermo Scientific Chemicals Supplier No. H52515MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 652148-91-9 | C5H5BClNO2 | 157.36 g/mol

5-Chloropyridine-2-boronic acid was used as reagent in organic synthesis. Also used as phamaceutical Intermediates. Used in suzuki-Miyaura coupling processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloropyridine-2-boronic acid was used as reagent in organic synthesis. Also used as phamaceutical Intermediates. Used in suzuki-Miyaura coupling processes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 652148-91-9
Molecular Formula C5H5BClNO2
Molecular Weight (g/mol) 157.36
MDL Number MFCD07368862
InChI Key JEGHCYRKSUGHJH-UHFFFAOYSA-N
Synonym 5-chloro-2-pyridineboronic acid, 5-chloropyridine-2-boronic acid, 5-chloropyridin-2-yl boronic acid, 2-borono-5-chloropyridine, 5-chloropyridine-2-boronicacid, 3-chloropyridine-6-boronic acid, 5-chloro-2-pyridyl boronic acid, boronic acid, 5-chloro-2-pyridinyl, pubchem13568, acmc-209npx
PubChem CID 22832086
IUPAC Name (5-chloropyridin-2-yl)boronic acid
SMILES B(C1=NC=C(C=C1)Cl)(O)O

Specifications

Quantity 250 mg
Formula Weight 157.37
Percent Purity 95%
Chemical Name or Material 5-Chloropyridine-2-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only